Learn More
Cayman Chemical 2-AcetylphenothIazIn 10g
SDP
Supplier:  Cayman Chemical 1905610 G
Encompass_Preferred
A selective, cell-active inhibitor of NOX1 that blocks the generation of ROS in HT-29 cells (IC50 = 0.129 µM); abolishes collagen-induced superoxide production by platelets (IC50 = 306 nM)
Catalog No. 50-298-1867