Learn More
Medchemexpress LLC 2-acetylthiomethyl-3-(4-methylbenzoyl) propionic acid | 101973-77-7 | MFCD00872973 | 99.9% | 280.34 g/mol | C14H16O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1944010MG
Encompass_Preferred
Esonarimod (KE-298) is a small-molecule antirheumatic research compound supplied as a dry powder for laboratory studies. It has been investigated for modulation of inflammatory responses and reached clinical evaluation for rheumatoid arthritis.
  • High purity of 99.9%.
  • Supplied as a solid powder suitable for formulation.
  • Soluble in DMSO at 100 mg/mL with ultrasonic assistance.
  • Long-term stability when stored at -20°C.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-94743