Learn More
Medchemexpress LLC 2-Acetylthiophene | 88-15-3 | 100.0% | 126.18 | 1 KG
SDP
Supplier:  Medchemexpress LLC HY205851KG
Encompass_Preferred
2-Acetylthiophene is a biochemical reagent used as a biological material or organic compound for life science research. It is described as a fragrance agent with a sulfurous odor and an onion-type flavor. It is suitable for various laboratory applications and research purposes, including those requiring specific solubility protocols.
  • Used as a biochemical reagent
  • Functions as a biological material or organic compound
  • Suitable for life science related research
  • Acts as a fragrance agent
  • Possesses a sulfurous type odor and an onion type flavor
Catalog No. 50-004-01862