Learn More
Medchemexpress LLC 2-Acetylthiophene | 88-15-3 | 99.8% | 126.18 | 25 G

Supplier: Medchemexpress LLC 25GHY20585
2-Acetylthiophene is a biochemical reagent suitable for life science-related research, functioning as a biological material or organic compound. It is also recognized as a fragrance agent due to its sulfurous odor and onion-like flavor.
- Appears as a colorless to light yellow liquid.
- Store pure form at -20°C for 3 years or 4°C for 2 years.
- Store in solvent at -80°C for 6 months or -20°C for 1 month.
- Soluble in DMSO up to 200 mg/mL.
- Available documentation includes Data Sheet, COA, SDS, Handling Instructions, HNMR, and GC.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.