Learn More
Medchemexpress LLC 2-Acetylthiophene | 88-15-3 | 99.84% | 126.18 | 100 G
Supplier: Medchemexpress LLC HY20585100G
2-Acetylthiophene is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is also described as a fragrance agent.
- Can be used as a biological material
- Functions as an organic compound for life science related research
- Acts as a fragrance agent
- Has a sulfurous type odor
- Provides an onion type flavor