Learn More
Medchemexpress LLC 2-Acetylthiophene | 88-15-3 | 99.98% | 126.18 | 500 G

Supplier: Medchemexpress LLC HY20585500G
2-Acetylthiophene is a biochemical reagent used as a biological material or organic compound for life science related research. It is also characterized as a fragrance agent with a sulfurous type odor and an onion type flavor.
- Appearance: Liquid
- Color: Colorless to light yellow
- Density: 1.1709 g/cm³
- Shipping: Room temperature in continental US
- Storage (pure form): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.