Learn More
Thermo Scientific Aluminum Rotor Blocks - Glass and Plastic Tubes, 13 x 100 mm
Supplier:  Thermo Scientific RBA3513100
Encompass_Preferred
13 x 100 mm. Holds 210 tubes (ESRG), 280 tubes (EXP-H200).
Catalog No. 50-870-559