Learn More
Medchemexpress LLC 2-Amino-1,3-bis(carboxylethoxy)propane hydrochloride | 2421153-68-4 | ≥97.0% | 271.70 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY23212A
Encompass_Preferred
2-Amino-1,3-bis(carboxylethoxy)propane hydrochloride is a non-protein amino acid and sulfamic acid. It contains one amino group and two terminal carboxylic acids. This compound has various physiological functions, including maintaining cell membrane stability, regulating cholesterol metabolism, and supporting nervous system function.
  • Participates in collagen synthesis
  • Acts as an anti-oxidant
  • Appearance: solid
  • Color: white to off-white
Catalog No. 50-004-02247