Learn More
Medchemexpress LLC 2-amino-1-(4-chlorobenzyl)-5-trifluoromethylbenzimidazole | 150493-34-8 | 99.8% | 325.72 g·mol⁻1 | C15H11ClF3N3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10142810MM 1ML
Encompass_Preferred
NS-638 is a small nonpeptide Ca2+-channel blocker used in research to inhibit K+-stimulated intracellular calcium elevation. It is commonly employed in in vitro pharmacology studies of voltage-gated calcium channels and calcium-dependent cellular responses.
  • Small nonpeptide Ca2+-channel blocker suitable for in vitro studies.
  • Blocks K+-stimulated intracellular Ca2+ elevation with IC50 = 3.4 μM.
  • Molecular formula C15H11ClF3N3; molecular weight 325.72 g·mol⁻1.
  • High reported purity (≈99.8%).
  • Available as a 10 mM solution in DMSO (1 mL) and in multiple solid weights for flexibility.
Catalog No. 50-002-49160