Learn More
Cayman Chemical 2-AmIno-1-phenylbutnhydro 10mg
SDP
Supplier:  Cayman Chemical 900186410 MG
Encompass_Preferred
An amphetamine characterized by having an ethyl group in the alpha position; partially substitutes for D-amphetamine in an animal discrimination analysis
Catalog No. 50-289-3626