Learn More
Medchemexpress LLC 2-Amino-1-phenylethanol | 7568-93-6 | 98.4% | C8H11NO | 1 ML

Supplier: Medchemexpress LLC HY5913210MM/1ML
2-Amino-1-phenylethanol is an analog of Norepinephrine and an endogenous metabolite. Its derivatives exhibit antimalarial activity against Plasmodium falciparum. This product is intended for research use only.
- Analog of Norepinephrine
- Derivatives show antimalarial activity against Plasmodium falciparum
- Endogenous metabolite
- Molecular weight of 137.18
- Appearance: solid, white to yellow
- Storage conditions: powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.