Learn More
Medchemexpress LLC 2-amino-2,3-dimethylbutanenitrile | 13893-53-3 | MFCD00661864 | 90.0% | 112.17 g/mol | C6H12N2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0408025G
Encompass_Preferred
AC-94149 is a substituted nitro compound used as an intermediate in herbicide synthesis and agrochemical research. It has the molecular formula C6H12N2, molecular weight 112.17 g/mol, CAS number 13893-53-3, and is typically supplied at 90.0% purity in small research pack sizes.
  • Used as an intermediate in herbicide synthesis and agrochemical research
  • Typical purity of 90.0% suitable for research applications
  • Available in small pack sizes for lab-scale work
  • Provided with datasheet, certificate of analysis, and safety data sheet
  • Molecular formula C6H12N2 and molecular weight 112.17 g/mol
Catalog No. 50-002-09107