Learn More
Medchemexpress LLC 2-Amino-2 -fluoro-5- 10g
SDP
Supplier:  Medchemexpress LLC HYW10407610G
Encompass_Preferred
2-Amino-2 -fluoro-5-nitrobenzophenone is a precursor in the synthesis of benzodiazepines 2-Amino-2 -fluoro-5-nitrobenzophenone is also an acid hydrolysis product of Flunitrazepam
Catalog No. 50-002-05239