Learn More
Medchemexpress LLC 2-Amino-2-methyl-1-propanol | 124-68-5 | 100.0% | 89.14 | 250 G
SDP
Supplier:  Medchemexpress LLC 250GHYW013027
Encompass_Preferred
2-Amino-2-methyl-1-propanol (AMP) functions as a neutralizer, buffer, and surfactant. It inhibits gelatinous product formation and is used for carbon dioxide absorption.
  • Functions as a neutralizer
  • Acts as a buffer
  • Serves as a surfactant
  • Inhibits gelatinous product formation
  • Used for carbon dioxide absorption
  • Suitable for histochemical demonstration of alkaline phosphatase activity (at 170-350 mM)
  • Does not cause liver effects in rats (at 300-380 mg/kg, dermal exposure)
  • Shows no compound-related changes in mice (at 0-600 ppm, p.o., 8 weeks)
Catalog No. 50-003-66346