Learn More
Medchemexpress LLC 2-Amino-2-methyl-1-propanol | 124-68-5 | 100.0% | 89.14 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW013027500G
Encompass_Preferred
2-Amino-2-methyl-1-propanol (AMP) is a versatile chemical acting as a neutralizer, buffer, and surfactant. It is notable for its ability to inhibit gelatinous product formation and is widely utilized for carbon dioxide absorption. This product is strictly intended for research purposes and is not suitable for patient use. Physically, it presents as a solid below 24°C and transforms into a liquid above 28°C, typically appearing colorless to light yellow.
  • Inhibits gelatinous product formation
  • Functions as a neutralizer, buffer, and surfactant
  • Utilized for carbon dioxide absorption
  • Intended for research use only
  • Appears as a solid below 24°C and liquid above 28°C
  • Colorless to light yellow appearance
Catalog No. 50-004-51198