Learn More
Medchemexpress LLC 2-Amino-2-methyl-1-propanol | 124-68-5 | 99.96% | 89.14 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013027100G
Encompass_Preferred
2-Amino-2-methyl-1-propanol (AMP) is a neutralizer, buffer, and surfactant. It inhibits gelatinous product formation and is used for carbon dioxide absorption. It is also used for histochemical demonstration of alkaline phosphatase activity. In animal studies, 2-amino-2-methyl-1-propanol (300-380 mg/kg, dermal exposure) does not cause liver effects in rats, and at 0-600 ppm (p.o., 8 weeks) shows no compound-related changes in mice. However, at 1100-1700 mg/kg (p.o., 30 days), it showed elevated SGPT and OCT serum levels in some rats, without accompanying gross pathology or histopathology changes.
  • Acts as a neutralizer, buffer, and surfactant
  • Inhibits gelatinous product formation
  • Used for carbon dioxide absorption
  • Used for histochemical demonstration of alkaline phosphatase activity
Catalog No. 50-004-53232