Learn More
Medchemexpress LLC 2-Amino-3-(2-chlorophenyl)propanoic acid | 14091-11-3 | 199.63 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00217010G
Encompass_Preferred
2-Amino-3-(2-chlorophenyl)propanoic acid is a phenylalanine derivative, recognized as a beneficial ergogenic dietary substance. It influences anabolic hormone secretion, provides fuel during exercise, and helps prevent exercise-induced muscle damage. This product is for research use only.
  • Phenylalanine derivative
  • Influences anabolic hormone secretion
  • Acts as fuel during exercise
  • Enhances mental performance under stress
  • Prevents exercise-induced muscle damage
Catalog No. 50-004-47845