Learn More
Medchemexpress LLC 2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid | 66-02-4 | MFCD00063076 | ≥95.0% | 432.98 g/mol | C9H9I2NO3 | 250 MG

Supplier: Medchemexpress LLC 250MGHYW052493
2-Amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid is an iodinated tyrosine derivative supplied for research use as a solid reagent. It is provided at analytical-grade purity for biochemical and analytical workflows requiring an iodinated amino acid.
- Iodinated tyrosine derivative for research use.
- Molecular formula C9H9I2NO3.
- Molecular weight 432.98 g/mol.
- Purity ≥95.0%.
- Available in 100 mg and 250 mg; larger quantities available by request.
- Store at 4°C and protect from light; in solvent, store at -80°C for long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.