Learn More
Medchemexpress LLC 2-amino-3',6'-bis(diethylamino)spiro[isoindole-3,9'-xanthene]-1-one | 74317-53-6 | MFCD07808762 | 99.9% | 456.58 g/mol | C28H32N4O2 | 25MG

Supplier: Medchemexpress LLC HY12364525MG
Rhodamine B hydrazide is a fluorescent derivative of rhodamine B used as a probe and labeling reagent for sensing metal ions and reactive oxygen/nitrogen species. It is supplied as an off-white to yellow solid with high analytical purity and strong visible-range fluorescence, and is typically handled and stored protected from light to preserve activity.
- High purity: 99.87%.
- Molecular weight: 456.58 g/mol.
- Chemical formula: C28H32N4O2.
- Solubility: soluble in DMSO at 100 mg/mL; ultrasonic agitation recommended.
- Applications: fluorescent probe for detection of Cu2+, Hg2+, peroxynitrite, hydroxyl radicals, and nitric oxide.
- Storage: store at 4°C protected from light; in solution, store at -80°C up to 6 months or -20°C up to 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.