Learn More
Medchemexpress LLC 2-amino-3',6'-bis(diethylamino)spiro[isoindole-3,9'-xanthene]-1-one | 74317-53-6 | MFCD07808762 | 99.9% | 456.58 g/mol | C28H32N4O2 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1236455MG
Encompass_Preferred
Rhodamine B hydrazide is a fluorescent derivative of rhodamine B used as a chemical probe in fluorescence-based assays to detect reactive species and certain metal ions. It is supplied as a light-sensitive solid with typical excitation near 510 nm and emission near 578 nm, and can convert to Rhodamine B upon reaction with sulfite. Recommended storage is refrigerated and protected from light; in solution, deep freezing extends stability.
Catalog No. 50-002-37707