Learn More
Medchemexpress LLC 2-Amino-3-benzyloxypyridine | 24016-03-3 | 99.9% | C12H12N2O | 5 G
SDP
Supplier:  Medchemexpress LLC HY752635G
Encompass_Preferred
2-Amino-3-benzyloxypyridine is a biochemical reagent that can be utilized as a biological material or organic compound for life science related research. It acts as an inhibitor of mitogen-activated protein kinase p38α activity.
  • Used as a biochemical reagent
  • Utilized as a biological material
  • Organic compound for life science related research
  • Inhibitor of mitogen-activated protein kinase p38α activity
Catalog No. 50-004-34378