Learn More
Medchemexpress LLC 2-Amino-3-hydroxypyridine | 16867-03-1 | 110.11 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007794
Encompass_Preferred
2-Amino-3-hydroxypyridine is a biochemical reagent used as a biological material or organic compound for life science-related research. This product is for research use only and is not intended for patient use.
  • Used as a biochemical reagent
  • Can be used as a biological material or organic compound
  • Suitable for life science related research
  • Functions as a chromogenic reagent for the spectrophotometric determination of osmium
Catalog No. 50-004-49863