Learn More
Medchemexpress LLC 2-Amino-3-hydroxypyridine | 16867-03-1 | 110.11 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW00779450G
Encompass_Preferred
2-Amino-3-hydroxypyridine is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is also suitable for the spectrophotometric determination of osmium.
  • Can be used as a biological material
  • Can be used as an organic compound for life science-related research
  • Can be used as a chromogenic reagent for the spectrophotometric determination of osmium
Catalog No. 50-004-48621