Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Amino-3-iodopyridine | 104830-06-0 | 99.9% | 220.01 | 5 G
SDP

Supplier:  Medchemexpress LLC HY328135G

Encompass_Preferred

2-Amino-3-iodopyridine (3-Iodopyridin-2-amine) can be coupled with formyl derivatives under a copper-catalyzed indole synthesis method, and then constructed azide analogs through copper-mediated conjugated adducts. The nucleophilicity of 2-Amino-3-iodopyridine is lower than that of the corresponding aniline used in the synthesis of indole, which presents a potential obstacle for future synthesis.

  • Solubility in DMSO: 200 mg/mL (requires ultrasonic)
  • Store at 4°C, protected from light
  • When in solvent, store at -80°C for 6 months; -20°C for 1 month (protected from light)
  • Targeted as biochemical assay reagents

Catalog No. 50-004-04035


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.