Learn More
Medchemexpress LLC 2-amino-3-methylpentan-1-ol (L-isoleucinol) | 24629-25-2 | MFCD00004731 | ≥97.0% | 117.19 g/mol | C6H15NO | 25 G

Supplier: Medchemexpress LLC 25GHYW015875
2-amino-3-methylpentan-1-ol (L-isoleucinol) is a chiral amino alcohol used as a biochemical reagent and building block in life-science research. It is employed in biochemical assays, stereoselective synthesis, and as an intermediate in medicinal chemistry and other laboratory workflows.
- Amino alcohol suitable for biochemical assays and organic synthesis.
- Chiral (2S,3S) stereochemistry for enantiomer-specific applications.
- Molecular formula C6H15NO; molecular weight 117.19 g/mol.
- Available in small lab pack sizes (10 g, 25 g, 50 g); larger quantities by quote.
- Store at 4°C and protect from light; in solvent: -80°C (up to 6 months), -20°C (up to 1 month).
- Product data sheet and SDS available for handling and safety information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.