Learn More
Medchemexpress LLC 2-amino-4-(4-chlorophenyl)thiazole | 2103-99-3 | MFCD00047058 | 99.8% | 210.69 | C9H7ClN2S | 5 G

Supplier: Medchemexpress LLC HYW0521905G
4-(4-chlorophenyl)thiazol-2-amine is an aminothiazole derivative supplied for laboratory research. It is provided as an off-white to light yellow solid with high purity suitable for analytical and synthetic applications.
- Cas number: 2103-99-3.
- Chemical formula: C9H7ClN2S.
- Molecular weight: 210.69.
- Purity: 99.8%.
- Appearance: off-white to light yellow solid.
- Typical use: analytical and synthetic research applications.
- Available packaging: 5 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.