Learn More
Medchemexpress LLC 2-Amino-4-bromo-3-nitropyridine | 84487-10-5 | 218.01 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW006690
Encompass_Preferred
2-Amino-4-bromo-3-nitropyridine is a drug intermediate primarily used for the synthesis of various active compounds. This product is intended for research use only and has not been fully validated for medical applications.
  • Used as a drug intermediate
  • Facilitates synthesis of various active compounds
  • Intended for research use only
Catalog No. 50-004-49926