Learn More
Medchemexpress LLC 2-Amino-4-chloropyridine | 19798-80-2 | MFCD04113820 | 99.9% | C5H5ClN2 | 100 G
SDP
Supplier:  Medchemexpress LLC HY60325100G
Encompass_Preferred
2-Amino-4-chloropyridine is a key intermediate in organic synthesis. Its derivatives demonstrate antibacterial activity against a range of microorganisms, including Gram-positive and Gram-negative bacteria, as well as fungi. This compound is also a useful precursor for synthesizing various chlorinated pyridine derivatives.
  • Key intermediate for organic synthesis
  • Derivatives show antibacterial activity against Gram-positive bacteria
  • Derivatives show antibacterial activity against Gram-negative bacteria
  • Derivatives show antibacterial activity against fungi
  • Useful precursor for synthesizing chlorinated pyridine derivatives
Catalog No. 50-004-34710