Learn More
Medchemexpress LLC 2-amino-4-pentynoic acid | 50428-03-0 | MFCD00056728 | 98.0% | 113.11 g·mol⁻¹ | C5H7NO2 | 100 MG

Supplier: Medchemexpress LLC HYW040124100MG
DL-Propargylglycine (2-amino-4-pentynoic acid, CAS 50428-03-0) is an alkyne-containing glycine derivative used as a click-chemistry reagent and research building block. It supports bioconjugation and small-molecule synthesis, and is supplied as a solid suitable for laboratory use.
- Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
- Useful as a small-molecule probe or building block in organic synthesis and bioconjugation.
- Supplied as a solid for accurate weighing and convenient storage.
- High purity suitable for research applications (98.0%).
- Molecular formula C5H7NO2; molecular weight 113.11 g·mol⁻¹.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.