Learn More
Medchemexpress LLC 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylic acid amide | 40106-12-5 | MFCD00622214 | 95.0% | 210.30 g/mol | C10H14N2OS | 100 MG

Supplier: Medchemexpress LLC HYW028350100MG
NSC727447 is a small-molecule research compound that inhibits ribonuclease H (RNase H) activity from HIV-1 and HIV-2. It has formula C10H14N2OS, a molecular weight of 210.30 g/mol, and is supplied with a reported purity of 95.0%. For research use only.
- Inhibits RNase H of HIV-1 and HIV-2 with low micromolar activity.
- Shows selectivity relative to bacterial and human RNase H.
- Molecular weight 210.30 g/mol and formula C10H14N2OS.
- Soluble in DMSO at 50 mg/mL; ultrasonic treatment may improve dissolution.
- Store solid at 4°C protected from light; aliquot solutions to avoid repeated freeze-thaw.
- Available in small package sizes suitable for laboratory research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.