Learn More
Medchemexpress LLC 2-amino-5-chlorobenzophenone | 719-59-5 | 99.9% | C13H10ClNO | 50 G
SDP
Supplier:  Medchemexpress LLC HY7778550G
Encompass_Preferred
2-Amino-5-chlorobenzophenone is a substituted benzophenone that can be utilized in the synthesis of benzodiazepines. Derivatives of 2-Amino-5-chlorobenzophenone have demonstrated skeletal muscle relaxation activity.
  • Used in the synthesis of benzodiazepines.
  • Derivative shows skeletal muscle relaxation activity.
  • Available in solid form.
  • High purity (99.98%).
Catalog No. 50-004-35020