Learn More
Medchemexpress LLC 2-Amino-5-chlorobenzophenone | 719-59-5 | 99.98% | C13H10ClNO | 100 G
SDP
Supplier:  Medchemexpress LLC HY77785100G
Encompass_Preferred
2-Amino-5-chlorobenzophenone is a substituted benzophenone that finds utility in various chemical syntheses. It is particularly noted for its role as a precursor in the synthesis of benzodiazepines, a class of psychoactive drugs. Derivatives of 2-Amino-5-chlorobenzophenone have also been observed to exhibit skeletal muscle relaxation activity.
  • Can be used in the synthesis of benzodiazepines
  • Derivatives demonstrate skeletal muscle relaxation activity
Catalog No. 50-004-33467