Learn More
Medchemexpress LLC 2-amino-6-[(1S,2R)-1,2,3-trihydroxypropyl]-3H-pteridin-4-one | 2009-64-5 | MFCD00075457 | 99.9% | 253.21 g/mol | C9H11N5O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW04005550MG
Encompass_Preferred
Neopterin is an analytical reference standard of D-(+)-neopterin (CAS 2009-64-5) used as a biomarker in immunomonitoring and inflammation research. It is provided as a high-purity crystalline powder intended for analytical assays, method development, and quantitative measurements in laboratories.
  • Analytical reference standard for D-(+)-neopterin.
  • High purity: 99.9% (HPLC).
  • Chemical formula C9H11N5O4; molecular weight 253.21 g/mol.
  • Suitable for biomarker, immunomonitoring, and inflammation research.
  • Available in multiple small-scale package sizes for laboratory use.
  • Provided with certificate of analysis supporting batch purity.
Catalog No. 50-003-01877