Learn More
Medchemexpress LLC 2-Amino-6-methylbenzoic acid | 4389-50-8 | 99.4% | 151.16 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY41496
Encompass_Preferred
2-Amino-6-methylbenzoic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Can be used as a biological material or organic compound
  • Reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d]oxazin-4-one
  • For research use only, not for sale to patients
Catalog No. 50-004-02392