Learn More
Medchemexpress LLC 2-amino-7-(2-furanyl)-7,8-dihydro-5(6H)-quinazolinone | 299442-43-6 | MFCD02026323 | 99.8% | 229.23 | C12H11N3O2 | 25 MG

Supplier: Medchemexpress LLC HY10319525MG
NKY80 is a selective, non-competitive inhibitor of adenylyl cyclase (AC) type V used in research to modulate AC catalytic activity in cardiac and pulmonary tissues. It is a non-nucleoside quinazolinone with reported activity against multiple AC isoforms.
- Selective non-competitive adenylyl cyclase type V inhibitor.
- Reported IC50: 8.3 μM (AC V), 132 μM (AC III), 1.7 mM (AC II).
- Molecular formula C12H11N3O2; molecular weight 229.23.
- Purity 99.76% as supplied.
- Soluble in DMSO; recommended powder storage at -20°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.