Learn More
Medchemexpress LLC 2-Amino-8-oxononanoic acid hydrochloride | 2984160-77-0 | 98.0% | C9H18ClNO3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY145576B50MG
Encompass_Preferred
2-Amino-8-oxononanoic acid (hydrochloride) is the hydrochloride form of 2-Amino-8-oxononanoic acid. This amino acid is incorporated into proteins in *E. coli* in genetic studies. It is efficient for labeling proteins with various probes in a site-specific manner under mild conditions, close to physiological pH.
  • Is the hydrochloride form of 2-amino-8-oxononanoic acid.
  • Is an amino acid.
  • Incorporated into proteins in *E. coli* in genetic studies.
  • Efficiently labels proteins with different probes.
  • Labels in a site-specific manner.
  • Functions under mild conditions close to physiological pH.
Catalog No. 50-003-56098