Learn More
Medchemexpress LLC 2-amino-n-(14-azido-2,5-dioxo-9,12-dioxa-3,6-diazatetradecyl)acetamide | 2222277-20-3 | 95.0% | 345.35 g/mol | C12H23N7O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15727510MM 1ML
Encompass_Preferred
Gly-Gly-Gly-PEG2-azide is an azide-functionalized peptide-PEG linker supplied as a ready-to-use solution (10 mM in DMSO, 1 mL) or as a solid. It is intended for use in antibody-drug conjugate synthesis, bioconjugation, and related chemical biology applications.
  • Azide-functionalized peptide-PEG2 linker for bioconjugation.
  • Available as 10 mM solution in DMSO (1 mL) or solid forms.
  • Used for antibody-drug conjugate synthesis and related chemistry.
  • Characterized by CAS 2222277-20-3 and molecular weight 345.35 g/mol.
Catalog No. 50-002-04483