Learn More
Medchemexpress LLC 2-amino-N-(4-chlorophenyl)benzamide | 4943-86-6 | MFCD00170586 | 98.9% | 246.69 g/mol | C13H11ClN2O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118639100MG
Encompass_Preferred
2-amino-N-(4-chlorophenyl)benzamide is a research-grade selective cyclooxygenase-1 (COX-1) inhibitor supplied as a solid for biochemical and pharmacological studies.
  • Selective cyclooxygenase-1 inhibitor suitable for COX-1 focused assays.
  • High purity (~98.9%) appropriate for analytical and biological testing.
  • Molecular weight 246.69 g/mol; formula C13H11ClN2O.
  • Solid form with recommended storage at 4°C and protection from light; solution stability: -80°C (6 months) or -20°C (1 month).
  • Available in multiple pack sizes, including 100 mg and solution aliquots for screening.
Catalog No. 50-002-29542