Learn More
Medchemexpress LLC 2-amino N-hydroxy-benzenecarboximidamide | 16348-49-5 | MFCD09910190 | 99.9% | 151.17 | C7H9N3O | 500MG

Supplier: Medchemexpress LLC HY125372500MG
2-Amino benzamidoxime is a small-molecule biochemical reagent used as a synthetic intermediate and reagent in biochemical assays. It is supplied as a solid and as a 10 mM solution in DMSO and is provided at high purity for research applications. Not for human or veterinary use.
- High purity: 99.85%.
- Molecular weight: 151.17.
- Chemical formula: C7H9N3O.
- Available pack sizes include 250 mg, 500 mg, 1 g, 5 g, 10 g, 50 g, and 10 mM in DMSO.
- Supplied as solid and as a solution for convenient use.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.