Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Aminobenzenesulfonamide | 3306-62-5 | 99.9% | 200 MG
SDP

Supplier:  Medchemexpress LLC HYB2147200MG

Encompass_Preferred

2-Aminobenzenesulfonamide is a carbonic anhydrase IX inhibitor. It is for research use only.

  • Appearance: Solid
  • Color: White to light brown
  • Molecular weight: 172.20
  • Formula: C6H8N2O2S
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 2 years, -20°C for 1 year

Catalog No. 50-004-11316


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.