Learn More
Medchemexpress LLC 2-aminobenzothiazole | 136-95-8 | MFCD00005785 | 99.7% | 150.21 g/mol | C7H6N2S | 100 G
SDP
Supplier:  Medchemexpress LLC HYW017424100G
Encompass_Preferred
2-Aminobenzothiazole (CAS 136-95-8) is a heterocyclic organic compound used as a building block in organic synthesis and as a ligand in coordination chemistry. Supplied for research use, it is available as a solid and as a DMSO solution for analytical and synthetic applications.
  • High purity (99.69%) suitable for research-grade applications.
  • Available in solid form and as a 10 mM solution in DMSO for flexible use.
  • Acts as a versatile building block for heterocyclic synthesis and derivatization.
  • Functions as a ligand in coordination and materials chemistry.
  • Compatible with common analytical techniques such as HPLC and NMR.
  • Identified by CAS number for traceability and regulatory use.
Catalog No. 50-002-99954