Learn More
Medchemexpress LLC 2-Aminoethyl diphenylborinate | 524-95-8 | 98.9% | 225.09 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW009724
Encompass_Preferred
2-Aminoethyl diphenylborinate (2-APB) is a cell-permeable inhibitor of Inositol triphosphate receptor (IP3R). It also inhibits the store-operated Ca2+ (SOC) channel and activates some TRP channels (V1, V2 and V3). Additionally, it has inhibitory effects on vasospasm. At high concentrations, it exhibits specific anti-inflammatory and antioxidant effects in neural tissue. This product is for research use only.
  • Inhibits inositol triphosphate receptor (IP3R)
  • Inhibits store-operated Ca2+ (SOC) channel
  • Activates some TRP channels (V1, V2, and V3)
  • Inhibitory effects on vasospasm
  • Exhibits specific anti-inflammatory and antioxidant effects in neural tissue (at high concentrations)
Catalog No. 50-004-53468