Learn More
Medchemexpress LLC 2-aminofluorene | 153-78-6 | MFCD00001125 | 99.9% | 181.24 g/mol | C13H11N | 1 G

Supplier: Medchemexpress LLC HYW0164331G
2-Aminofluorene is an aromatic amine research reagent (CAS 153-78-6) used as a genotoxin in studies of DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. It is supplied as a powder for laboratory research; follow recommended storage conditions for solids and solutions.
- High purity (99.9%) suitable for analytical and biological studies.
- Physical form: powder, convenient for weighing and dissolving.
- Intended for genotoxicity research, including DNA adduct formation and DNA repair studies.
- Storage recommendations: powder at -20°C (up to 3 years); solutions at -80°C for long-term storage.
- Molecular formula C13H11N; molecular weight 181.24 g/mol.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.