Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-aminofluorene | 153-78-6 | MFCD00001125 | 99.9% | 181.24 g/mol | C13H11N | 1 ML
SDP

Supplier:  Medchemexpress LLC HYW01643310MM/1ML

Encompass_Preferred

2-Aminofluorene (CAS 153-78-6) is an aromatic amine genotoxin used in molecular biology research to investigate DNA adduct formation, DNA repair, carcinogenesis, and mutagenesis. Available in both ready-to-use solution and solid formats, the material is provided for laboratory research applications only.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High stated purity, 99.9%.
  • Molecular formula C13H11N; molecular weight 181.24 g/mol.
  • Suitable for studies of DNA adducts, DNA repair, carcinogenesis, and mutagenesis.
  • Supplied as solids in multiple gram sizes for scale-up or bulk use.
  • For research use only; not for human or veterinary use.

Catalog No. 50-003-02198


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.