Learn More
Medchemexpress LLC 2-aminofluorene-d11 | 347841-44-5 | 99.0% | 192.3 g/mol | C13D11N | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW016433S110MG
Encompass_Preferred
2-Aminofluorene-d11 is the deuterium-labeled analogue of 2-aminofluorene used as an isotopically labeled internal standard and tracer in analytical and research workflows. It has chemical formula C13D11N, molecular weight 192.3 g/mol, and is associated with CAS number 347841-44-5.
  • Deuterium-labeled (d11) for accurate isotope-dilution quantitation.
  • High chemical purity (99.0% by HPLC) for analytical reliability.
  • High isotopic enrichment (~98% D) to ensure a clear mass shift in MS analyses.
  • Available in small milligram quantities suitable for method development and internal standard use.
  • Suitable for LC-MS/MS, isotope-dilution assays, and tracer studies.
Catalog No. 50-002-99343