Learn More
Selleck Chemical LLC 2-Aminoisobutyric acid
SDP
Supplier:  Selleck Chemical LLC S627025MG
Encompass_Preferred
2-Aminoisobutyric acid (NSC-16590 2-Methylalanine H-Aib-OH) is a rare non-protein amino acid and end-product of pyrimidine metabolism excreted in urine and found in some antibiotics of fungal origin
Catalog No. 50-000-03175