Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-Aminomethylpyrimidine (hydrochloride) | 372118-67-7 | 99.91% | C5H8ClN3 | 50 G
SDP

Supplier:  Medchemexpress LLC HY60347A50G

Encompass_Preferred

2-Aminomethylpyrimidine hydrochloride (Pyrimidin-2-ylmethanamine hydrochloride) is a synthetic intermediate useful for pharmaceutical synthesis. It is an off-white to light yellow solid with a molecular weight of 145.59, and typically has a purity of 99.91%.

  • Useful for pharmaceutical synthesis
  • For research use only
  • Off-white to light yellow solid
  • Purity of 99.91%
  • Store at room temperature (RT) in a sealed container, away from moisture and light
  • Store in solvent at -80°C for 1 year or -20°C for 6 months

Catalog No. 50-004-34124


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.