Learn More
Medchemexpress LLC 2-aminopurine-o-ph-nhco-c3-cooh hydrochloride | 95.3% | 420.85 | C18H21ClN6O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW422359A10MG
Encompass_Preferred
2-Aminopurine-O-Ph-NHCO-C3-COOH hydrochloride is the acid-form intermediate used in the synthesis of the corresponding NHS ester. Supplied as the hydrochloride salt, it is a white to off-white solid with a reported purity of 95.3% and molecular formula C18H21ClN6O4. It is intended for use as a chemical intermediate in research and synthetic workflows and should be stored sealed and away from moisture.
  • Hydrochloride salt form for improved stability.
  • White to off-white solid for easy handling and weighing.
  • Reported purity 95.3% suitable for synthetic applications.
  • Molecular formula C18H21ClN6O4; molecular weight 420.85.
  • Recommended sealed storage; in solvent: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-002-81180