Learn More
Medchemexpress LLC 2-Aminoquinoline | 580-22-3 | 144.17 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00752410MM/1ML
Encompass_Preferred
2-Aminoquinoline (2-Quinolinamine) is a promising compound as a bioavailable nNOS inhibitor. It exhibits antiviral activity against the vaccinia virus and has potential for the research of antineurodegenerative agents. However, it suffers from low human nNOS inhibition, low selectivity versus human eNOS, and significant binding to other CNS targets.
  • Bioavailable nNOS inhibitor
  • Antiviral activity against vaccinia virus
  • Potential for antineurodegenerative agent research
  • Low human nNOS inhibition
  • Low selectivity versus human eNOS
  • Significant binding to other CNS targets
Catalog No. 50-004-48696