Learn More
Medchemexpress LLC 2-Aminoquinoline | 580-22-3 | 144.17 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW007524100G
Encompass_Preferred
2-Aminoquinoline is a promising compound as a bioavailable nNOS inhibitor, despite exhibiting low human nNOS inhibition, low selectivity versus human eNOS, and significant binding to other CNS targets. It demonstrates antiviral activity against the vaccinia virus and has potential for research into antineurodegenerative agents. This product is for research use only.
  • Antiviral activity against vaccinia virus.
  • Potential for research into antineurodegenerative agents.
  • For research use only.
Catalog No. 50-004-45806